missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tiotidine

Description
Potent histamine H2-receptor antagonist with negligible activity against H1- and H3- receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 312.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 312.414 |
| Chemical Name or Material | Tiotidine |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?