missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Antibody (2116C) - Azide and BSA Free, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB7442-SP
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Blocking/Neutralizing | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Blocking Assay | |
| 2116C | |
| Unconjugated | |
| PBS with No Preservative | |
| TNFSF15 | |
| Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150] | |
| 25 μg | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 9966 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction