missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Antibody (2116C) - Azide and BSA Free, R&D Systems™
Rabbit Monoclonal Antibody
194.00€ - 546.00€
Specifications
| Antigen | TL1A/TNFSF15 |
|---|---|
| Clone | 2116C |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Blocking/Neutralizing |
| Applications | Western Blot, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18063634
|
Novus Biologicals
MAB7442-SP |
25 μg |
194.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014596
|
Novus Biologicals
MAB7442 |
100 μg |
546.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
| TL1A/TNFSF15 | |
| 1.0 mg/mL | |
| Western Blot, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| PBS with No Preservative | |
| 9966 | |
| Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 2116C | |
| Western Blot, Blocking/Neutralizing | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| TNFSF15 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title