missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB100-56588SS
This item is not returnable.
View return policy
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).
Specifications
| TLR7 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| toll-like receptor 7 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Reacts with Human and Mouse. 72% sequence identity with rat protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen 1:20. Use 1:20, Flow (Cell Surface) reported in scientific literature (Wong et al (2007)), Flow (Intracellular) 0.5 - 2 ug/1x10^6 cells | |
| Q9NYK1 | |
| TLR7 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 684-701 of human TLR7 (NP_057646). | |
| 0.025 mg | |
| Immunology, Innate Immunity, Toll Like Receptors | |
| 51284 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction