missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
226.00€ - 476.00€
Specifications
| Antigen | TLR7 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen 1:20. Use 1:20, Flow (Cell Surface) reported in scientific literature (Wong et al (2007)), Flow (Intracellular) 0.5 - 2 ug/1x10^6 cells |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038585
|
Novus Biologicals
NB100-56588SS |
0.025 mg |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767713
|
Novus Biologicals
NB100-56588 |
0.1 mg |
476.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).Specifications
| TLR7 | |
| Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen 1:20. Use 1:20, Flow (Cell Surface) reported in scientific literature (Wong et al (2007)), Flow (Intracellular) 0.5 - 2 ug/1x10^6 cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q9NYK1 | |
| 51284 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 684-701 of human TLR7 (NP_057646). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Toll Like Receptors | |
| PBS with 0.05% Sodium Azide | |
| toll-like receptor 7 | |
| TLR7 | |
| IgG | |
| Immunogen affinity purified | |
| Reacts with Human and Mouse. 72% sequence identity with rat protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title