Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand:  Novus Biologicals NB100-56588
 View more versions of this product
Product Code. 18767713
  • 476.00€ / 0.10mg
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR7
Rabbit
RUO
Immunohistochemistry
0.1 mg