Learn More
Invitrogen™ TMCO5A Polyclonal Antibody
GREENER_CHOICE
Product Code. 17131084 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17131084 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17131084 Supplier Invitrogen™ Supplier No. PA5113222
Rabbit Polyclonal Antibody
TMCO5A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen TMCO5A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMCO5A
Gene Accession No. Q8N6Q1
Gene Alias testicular tissue protein Li 205; TMCO5; TMCO5A; Transmembrane and coiled-coil domain-containing protein 5A; transmembrane and coiled-coil domains 5; transmembrane and coiled-coil domains 5A
Gene Symbols TMCO5A
Host Species Rabbit
Immunogen Recombinant Human Transmembrane and coiled-coil domain-containing protein 5A protein (10-192AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145942
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG