missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPCN1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16587981
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16587981 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16587981 Supplier Invitrogen™ Supplier No. PA590758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FRMNYSRKNQ DSEVDGGITL EKEISKEELV AVLELYREAR GASSDVTRLL ETLSQMERYQ QHSMVFLGRR SRTKSDLSLK MYQEEIQEWY EEHAREQEQQ RQLSSSAAPA AQQPPGSRQR SQTVT; Positive Samples: Rat lung; Cellular Location: Endosome membrane, Lysosome membrane, Multi-pass membrane protein.

TPCN1 may function as one of the major voltage-gated Ca2+ channel (VDCC) across the plasma membrane. Voltage-gated Ca(2+) and Na+ channels have 4 homologous domains, each containing 6 transmembrane segments, S1 to S6. TPCN1 is similar to these channels, but it has only 2 domains containing S1 to S6 (Ishibashi et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPCN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TPCN1
Gene Accession No. Q9EQJ0, Q9WTN5
Gene Alias 5730403B01Rik; FLJ20612; Kiaa1169; mKIAA1169; si:dkey-125i10.5; TPC1; TPCN1; Two pore calcium channel protein 1; two pore channel 1; two pore segment channel 1; two-pore channel 1; two-pore channel 1, homolog; two-pore segment channel 1; voltage-dependent calcium channel protein TPC1; voltage-gated Ca channel
Gene Symbols TPCN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 692-816 of human TPCN1 (NP_0603712).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246215, 252972
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.