missing translation for 'onlineSavingsMsg'
Learn More

Tralkoxidym, TRC

Product Code. 17943385
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943385 500 mg 500mg
17944645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17943385 Supplier Toronto Research Chemicals Supplier No. TRCT712403500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87820-88-0
Molecular Formula C20 H27 N O3
Molecular Weight (g/mol) 329.43
Synonym 2-[1-(Ethoxyimino)propyl]-3-hydroxy-5-(2,4,6-trimethylphenyl)-2-cyclohexen-1-one (ACI), 2-[1-(Ethoxyimino)propyl]-3-hydroxy-5-mesitylcyclohex-2-enone, Achieve, Achieve (pesticide), FD 4026, Grasp, Grasp (cyclohexenone), ICI-A 604, ICIA 0604, PP 604, Splendor, Tralkoxydim
IUPAC Name 2-[(E)-N-ethoxy-C-ethylcarbonimidoyl]-3-hydroxy-5-(2,4,6-trimethylphenyl)cyclohex-2-en-1-one
SMILES CCO\N=C(/CC)\C1=C(O)CC(CC1=O)c2c(C)cc(C)cc2C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H27NO3/c1-6-16(21-24-7-2)20-17(22)10-15(11-18(20)23)19-13(4)8-12(3)9-14(19)5/h8-9,15,22H,6-7,10-11H2,1-5H3/b21-16+
Formula Weight 329.1991
Chemical Name or Material Tralkoxydim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Política de privacidad.