missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid, 98%, Thermo Scientific™

Product Code. 10572461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572461 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572461 Supplier Thermo Scientific Acros Supplier No. 298690010
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 212757-09-0
Melting Point 163°C to 169°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H15O3Br
MDL Number MFCD00800528
Quantity 1 g
Synonym 1r,2r-2-4-bromobenzoyl cyclohexane-1-carboxylic acid, trans-2-4-bromobenzoyl-1-cyclohexane-carboxylic acid, trans-2-4-bromobenzoyl cyclohexanecarboxylic acid, trans-2-4-bromobenzoyl-1-cyclohexane-carboxylic acid, 98, 1r,2r-2-4-bromophenyl carbonyl cyclohexanecarboxylic acid, 1r,2r-2-4-bromobenzoyl cyclohexanecarboxylic acid, cyclohexanecarboxylicacid, 2-4-bromobenzoyl-, 1r,2r-rel
InChI Key OVZXXISOLDHARA-VXGBXAGGSA-N
SMILES C1CCC(C(C1)C(=O)C2=CC=C(C=C2)Br)C(=O)O
IUPAC Name (1R,2R)-2-(4-bromobenzoyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 311.17
PubChem CID 2725095
Formula Weight 311.17
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.