missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-2-butenal, 97%

Product Code. 11397985
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397985 25 g 25g
11387985 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397985 Supplier Thermo Scientific Alfa Aesar Supplier No. L08113.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 497-03-0 | C5H8O | 84.118 g/mol

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Density 0.871
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Odor Fruit-like
Refractive Index 1.448
Quantity 25 g
UN Number UN1989
Beilstein 1698207
Sensitivity Air sensitive
Solubility Information Soluble in water
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-butenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.