missing translation for 'onlineSavingsMsg'
Learn More

TREML2/TLT-2 Antibody (001), Biotin , Novus Biologicals™

Product Code. 18666651 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18666651 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18666651 Supplier Novus Biologicals Supplier No. NBP289948B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TREML2/TLT-2 Monoclonal antibody specifically detects TREML2/TLT-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREML2/TLT-2
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate Biotin
Formulation PBS
Gene Alias C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TREML2/TLT-2 (NP_079083.2; Met1-Ser268).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79865
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.