missing translation for 'onlineSavingsMsg'
Learn More

Tri(4-morpholinyl)phosphine oxide, 99%

Product Code. 11362987
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362987 25 g 25g
11352987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362987 Supplier Thermo Scientific Alfa Aesar Supplier No. L16315.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4441-12-7 | C24H42 | 330.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4441-12-7
MDL Number MFCD00047406

Specifications

Melting Point 188°C to 192°C
Molecular Formula C12H24N3O4P
Quantity 25 g
Beilstein 280155
Synonym trimorpholinophosphine oxide, 4,4',4-phosphoryltrimorpholine, tri 4-morpholinyl phosphine oxide, phosphoric acid trimorpholide, 4-bis morpholin-4-yl phosphoroso morpholine, tmpo;tris-morpholino-phosphine oxide, phosphoric trimorpholide, 5-n-propylbenzoic acid, acmc-2097fl, ksc493m7l
InChI Key WXMQHPKQCPCDQO-UHFFFAOYSA-N
SMILES C1COCCN1P(=O)(N2CCOCC2)N3CCOCC3
IUPAC Name 4-dimorpholin-4-ylphosphorylmorpholine
Molecular Weight (g/mol) 305.32
PubChem CID 78180
Formula Weight 305.32
Percent Purity 99%
Chemical Name or Material Tri(4-morpholinyl)phosphine oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.