missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylallylphosphonium Bromide-d5, TRC

Product Code. 30456594
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456594 5 mg 5mg
30453758 10 mg 10mg
30457285 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456594 Supplier Toronto Research Chemicals Supplier No. TRCT7732425MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tri-n-butylallylphosphonium Bromide-d5
Molecular Formula C15H27D5BrP
Quantity 5 mg
Synonym Allyltri-n-butylphosphonium Bromide-d5, NSC 222446-d5, Tributyl-2-propenylphosphonium Bromide-d5, Allyltributylphosphonium Bromide-d5, Tributyl-2-propen-1-yl-phosphonium Bromide-d5
InChI Formula InChI=1S/C15H32P.BrH/c1-5-9-13-16(12-8-4,14-10-6-2)15-11-7-3;/h8H,4-7,9-15H2,1-3H3;1H/q+1;/p-1/i4D2,8D,12D2;
SMILES CCCC[P+](CCCC)(CCCC)C([2H])([2H])/C([2H])=C([2H])\[2H].[Br-]
IUPAC Name (allyl-d5)tributylphosphonium bromide
Molecular Weight (g/mol) 328.32
Formula Weight 327.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.