missing translation for 'onlineSavingsMsg'
Learn More

Triazophos 85%, TRC

Product Code. 30455074
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455074 1 g 1g
30453315 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455074 Supplier Toronto Research Chemicals Supplier No. TRCT7675951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24017-47-8
Molecular Formula C12 H16 N3 O3 P S
Molecular Weight (g/mol) 313.31
Synonym Phosphorothioic acid, O,O-diethyl O-(1-phenyl-1H-1,2,4-triazol-3-yl) ester, 1H-1,2,4-Triazol-3-ol, 1-phenyl-, O-ester with O,O-diethyl phosphorothioate (8CI), 1-Phenyl-3-(O,O-diethylthionophosphoryl)-1,2,4-triazole, HOE 2960, Hostathion, Hostathion 40EC, Hostation, Triazofos, Triazophos
IUPAC Name diethoxy-[(1-phenyl-1,2,4-triazol-3-yl)oxy]-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1ncn(n1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3
Formula Weight 313.065
Chemical Name or Material Triazophos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.