accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24017-47-8

IUPAC Name: O,O-diethyl O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate
Synonyms: triazophos hostathion triazofosz (hungarian) methoxone triazofos
Molecular Weight (g/mol): 313.31
Molecular Formula: C12H16N3O3PS
InChi Key: AMFGTOFWMRQMEM-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=NN(C=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Concentration 
  • (2)
  • (2)
  • (1)
Solvent or Matrix 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)