Learn More
Triclosan Methyl-d3 Ether, CDN
Product Code. 30315972
Change view
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315972 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315972 Supplier CDN Supplier No. CDND70740.05G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TriclosanmethylD3(methoxyD3)
CAS 1020720-00-6
Molecular Formula C13 2H3 H6 Cl3 O2
Quantity 0.05 g
Synonym Benzene, 4-chloro-1-(2,4-dichlorophenoxy)-2-(methoxy-d3)-, Triclosan methyl D3 (methoxy D3), Triclosan Methyl-d3 Ether
InChI Formula InChI=1 S/C13H9Cl3O2/c1-17-13-7-9(15)3-5-12(13)18-11-4-2-8(14)6-10(11)16/h2-7 H,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl
IUPAC Name 2,4-dichloro-1-[4-chloro-2-(trideuteriomethoxy)phenoxy]benzene
Molecular Weight (g/mol) 306.59
Formula Weight 304.9856 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity