missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthopropionate, 97%

Product Code. 11318025
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318025 500 mL 500mL
11398015 25 mL 25mL
11308025 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318025 Supplier Thermo Scientific Alfa Aesar Supplier No. L08172.AP
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 115-80-0 | C9H20O3 | 176.26 g/mol

Used as intermediates in pharmaceutical chemical and organic synthesize. It can react with N-benzoyl-glycine in the presence of 4-(dimethylamino)pyridine and acetic anhydride to produce 4-(1-ethoxy-propylidene)-2-phenyl-4H-oxazol-5-one. It is also applied as a starting material in the synthesis of Triethyl orthoacrylate. Triethyl orthopropionate is also commonly used in Claisen rearrangement reactions that yield γ,δ-unsaturated esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediates in pharmaceutical chemical and organic synthesize. It can react with N-benzoyl-glycine in the presence of 4-(dimethylamino)pyridine and acetic anhydride to produce 4-(1-ethoxy-propylidene)-2-phenyl-4H-oxazol-5-one. It is also applied as a starting material in the synthesis of Triethyl orthoacrylate. Triethyl orthopropionate is also commonly used in Claisen rearrangement reactions that yield γ,δ-unsaturated esters.

Solubility
Hydrolyzes slowly in water. Soluble in alcohol, chloroform, ethyl acetate and ether.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect against electrostatic charges. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-80-0
Molecular Formula C9H20O3
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009226
InChI Key FGWYWKIOMUZSQF-UHFFFAOYSA-N
Synonym triethyl orthopropionate, ethyl orthopropionate, propane, 1,1,1-triethoxy, triethylorthopropionate, triethyl o-propionate, orthopropionic acid ethyl ester, orthopropionic acid, triethyl ester, unii-9uvm48bas9, 9uvm48bas9, 1,1,1-triethoxypropan
PubChem CID 66995
IUPAC Name 1,1,1-triethoxypropane
SMILES CCOC(CC)(OCC)OCC

Specifications

Density 0.88
Boiling Point 158°C
Flash Point 45°C (113°F)
Odor Slightly Sweetish
Linear Formula CH3CH2C(OCH2CH3)3
Refractive Index 1.402
Quantity 500 mL
UN Number UN3272
Beilstein 906798
Solubility Information Hydrolyzes slowly in water. Soluble in alcohol,chloroform,ethyl acetate and ether.
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material Triethyl orthopropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.