missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, TRC

Product Code. 30041041
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041041 1 g 1g
30003449 10 g 10g
30007076 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041041 Supplier Toronto Research Chemicals Supplier No. TRCT7757401G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6 H15 N
Molecular Weight (g/mol) 101.19
Synonym Triethylamine (7CI,8CI), N,N-Diethylethanamine, (Diethylamino)ethane, LE 11-5RG, N,N,N-Triethylamine, TEA
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3
Formula Weight 101.1204
Chemical Name or Material Triethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.