missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylphosphine, 98

Product Code. 10263032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10263032 1 g 1g
10662983 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10263032 Supplier Thermo Scientific Acros Supplier No. 317330010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6476-36-4 | C9;H2;1;P | 160.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6476-36-4
Molecular Formula C9;H2;1;P
Molecular Weight (g/mol) 160.24

Specifications

Density 0.8390 g/mL
Boiling Point 81.0°C (22.0 mmHg)
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4690 to 1.4710
Quantity 1 g
Specific Gravity 0.8390
Formula Weight 160.24
Chemical Name or Material Triisopropylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.