missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM34 Monoclonal Antibody (K24023_20A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556190
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: GEKLLLFCKE DRKVICWLCE RSQEHRGHHT VLTEEVFKEC QEKLQAVLKR LKKEEEEAEK LEADIREEKT SWKYQVQTER QRIQTEFDQL RSILNNEEQR ELQRLEEEEK KTLDKFAEAE
TRIM34 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Expression of this gene is up-regulated by interferon. This gene is mappped to chromosome 11p15, where it resides within a TRIM gene cluster. Alternate splicing of this gene generates four transcript variants. Additionally, a read-through transcript transcribed from this gene and TRIM6 has been observed.
Specifications
| TRIM34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9BYJ4 | |
| TRIM34 | |
| Recombinant human TRIM34 (immunogen range is 101-220). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K24023_20A3 | |
| Unconjugated | |
| TRIM34 | |
| IFP1; Interferon-responsive finger protein 1; RING finger protein 21; ring finger protein 21, interferon-responsive; RNF21; TRIM34; tripartite motif containing 34; tripartite motif-containing 34; tripartite motif-containing protein 34 | |
| Mouse | |
| Protein A | |
| RUO | |
| 53840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction