missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM34 Monoclonal Antibody (K24023_20A3), Invitrogen™
Mouse Monoclonal Antibody
480.00€ - 846.00€
Specifications
| Antigen | TRIM34 |
|---|---|
| Clone | K24023_20A3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30195575
|
Invitrogen™
MA555810 |
50 μL |
480.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211672
|
Invitrogen™
MA556190 |
200 μL |
846.00€
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GEKLLLFCKE DRKVICWLCE RSQEHRGHHT VLTEEVFKEC QEKLQAVLKR LKKEEEEAEK LEADIREEKT SWKYQVQTER QRIQTEFDQL RSILNNEEQR ELQRLEEEEK KTLDKFAEAE
TRIM34 Monoclonal antibody specifically detects TRIM34 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| TRIM34 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| IFP1; Interferon-responsive finger protein 1; RING finger protein 21; ring finger protein 21, interferon-responsive; RNF21; TRIM34; tripartite motif containing 34; tripartite motif-containing 34; tripartite motif-containing protein 34 | |
| Recombinant human TRIM34 (immunogen range is 101-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TRIM34 |
| K24023_20A3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BYJ4 | |
| 53840 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title