missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthoacetate, 98%

Product Code. 11308086
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308086 50 mL 50mL
11318086 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308086 Supplier Thermo Scientific Alfa Aesar Supplier No. L13369.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1445-45-0 | C5H12O3 | 120.148 g/mol

Trimethyl orthoacetate is used as a diols can be protected as their 1-methoxyethylidene cyclic acetals (2-methoxy-2-methyl-1,3-dioxolanes) by acid-catalyzed exchange. It is mainly used as the chemical intermediates of the medicine. Some of the other applications includes it is mainly used as chemical intermediate of pharmaceutical and agricultural chemicals, used for compound medical intermediates of vitamin B1, vitamin A1 and sulfanilamide organism, used for dye and spices industry, Medicine, agricultural chemicals, paint additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl orthoacetate is used as a diols can be protected as their 1-methoxyethylidene cyclic acetals (2-methoxy-2-methyl-1,3-dioxolanes) by acid-catalyzed exchange. It is mainly used as the chemical intermediates of the medicine. Some of the other applications includes it is mainly used as chemical intermediate of pharmaceutical and agricultural chemicals, used for compound medical intermediates of vitamin B1, vitamin A1 and sulfanilamide organism, used for dye and spices industry, Medicine, agricultural chemicals, paint additives.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-45-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.148
MDL Number MFCD00008477
InChI Key HDPNBNXLBDFELL-UHFFFAOYSA-N
Synonym trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate
PubChem CID 15050
IUPAC Name 1,1,1-trimethoxyethane
SMILES CC(OC)(OC)OC

Specifications

Density 0.956
Boiling Point 107°C to 109°C
Flash Point 16°C (60°F)
Linear Formula CH3C(OCH3)3
Refractive Index 1.389
Quantity 50 mL
UN Number UN3272
Beilstein 1098338
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Trimethyl orthoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.