Learn More
Trimethyl orthoacetate, 98+%
Product Code. 10647022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647022 500 mL Glass bottle 500mL
10308070 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10647022 Supplier Thermo Scientific Acros Supplier No. 218185000
  • ONLINE EXCLUSIVE PRICE
    225.00€
    194.65€ / 500mL
    Save 30.35€
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 1445-45-0 | C5H12O3 | 120.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1445-45-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00008477
InChI Key HDPNBNXLBDFELL-UHFFFAOYSA-N
Synonym trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate
PubChem CID 15050
IUPAC Name 1,1,1-trimethoxyethane
SMILES CC(OC)(OC)OC
Specifications
Color Colorless to Brown
Density 0.9400g/mL
Boiling Point 107.0°C to 109.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(OCH3)3
Refractive Index 1.3870 to 1.3890
Quantity 500 mL
Beilstein 02, II, 128
Fieser 17,377; 01,834; 02,318
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolysis. Other solubilities: forming methanol and acetic acid
Formula Weight 120.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Trimethyl orthoacetate, 98%