missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMU Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 572.00€
Specifications
| Antigen | TRMU |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642428
|
Novus Biologicals
NBP2-68574-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667818
|
Novus Biologicals
NBP2-68574 |
100 μg |
572.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRMU Polyclonal antibody specifically detects TRMU in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TRMU | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 2.8.1, EC 2.8.1.-, FLJ10140, mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase, mitochondrial tRNA-specific 2-thiouridylase 1, MTO2 homolog, MTO2MGC99627, MTU1, TRMT, TRMT1, tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase, tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase, TRNT1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EDNKVLGTHKGWFLYTLGQRANIGGLREPWYVVEKDSVKGDVFVAPRTDHPALYRDLLRTSRVHWIAEEPPAALVRDKMMECHFRFRHQMALVPCVLTLNQDGTVWVTAVQAVRALATGQFAVFYKGDECLGS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 55687 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title