missing translation for 'onlineSavingsMsg'
Learn More

TRMU Monoclonal Antibody (OTI2B6), TrueMAB™, OriGene

Product Code. 17554512
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17554512 30 μL 30µL
17534512 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17554512 Supplier Origene Technologies Supplier No. TA505697S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRMU Monoclonal antibody specifically detects TRMU in Human samples. It is validated for Western Blot

Specifications

Antigen TRMU
Applications Western Blot
Classification Monoclonal
Clone OTI2B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ARPC5
Gene Accession No. O15511
Gene Alias 1110005N20Rik; 1600025P05Rik; AI314320; LCAL3; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; mitochondrial tRNA-specific 2-thiouridylase 1; MTO2; MTO2 homolog; MTU1; TRMT; Trmt1; Trmu; tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase 1; tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase; tRNA mitochondrial 2-thiouridylase; TRNT1
Host Species Mouse
Immunogen Full length human recombinant protein of human ARPC5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10092
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.