missing translation for 'onlineSavingsMsg'
Learn More

Tryptase epsilon/BSSP-4/PRSS22 Antibody (725445) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30614930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30614930 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30614930 Supplier Novus Biologicals Supplier No. FAB5047J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tryptase epsilon/BSSP-4/PRSS22 Monoclonal antibody specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tryptase epsilon/BSSP-4/PRSS22
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 725445
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase epsilon /BSSP-4, Ala33-Ser317, Accession # Q9GZN4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 64063
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.