Learn More
Invitrogen™ TPH1 Polyclonal Antibody
Description
PA1-777 detects TPH1 in human, mouse, rat, rabbit and non-human primate samples. PA1-777 has been successfully used in ICC/IF, IP, and Western blot procedures. By Western blot PA1-777 detects a 51 kDa protein representing TPH1. The PA1-777 immunogen is a synthetic peptide corresponding to the C-terminal region of TPH1.
Specifications
Specifications
| Antigen | TPH1 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | TPH1 |
| Gene Accession No. | P09810, P17290, P17532, P17752 |
| Gene Alias | indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; MGC119994; TPH; Tph1; TPH2; TPRH; TRPH; tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase; tryptophan hydroxylase (tryptophan 5-monooxygenase); tryptophan hydroxylase 1; tryptophan hydroxylase 1 (tryptophan 5-monooxygenase) |
| Gene Symbols | TPH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.