missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: FLANAKESPQ EEEIDPFDVD SGREFGNPNR PVASTRLPSD TDDSDASEDP Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 74%.
TSR3 is a probable pre-rRNA processing protein involved in ribosome biogenesis.
Specifications
Specifications
| Antigen | TSR3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TSR3 |
| Gene Accession No. | Q9UJK0 |
| Gene Alias | C16orf42; MGC24381; probable ribosome biogenesis protein C16orf42; ribosome biogenesis protein TSR3 homolog; TSR3; TSR3, 20S rRNA accumulation, homolog; TSR3, acp transferase ribosome maturation factor; UND313L |
| Gene Symbols | TSR3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
