missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Two pore calcium channel protein 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30670
This item is not returnable.
View return policy
Description
Two pore calcium channel protein 2 Polyclonal specifically detects Two pore calcium channel protein 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Two pore calcium channel protein 2 | |
| Polyclonal | |
| Unconjugated | |
| Sterilized water with No Preservative | |
| FLJ41094, SHEP10, TPC2two pore calcium channel protein 2, two pore segment channel 2, two-pore calcium channel protein 2, Voltage-dependent calcium channel protein TPC2 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 219931 | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 10-50 μg/mL, Immunohistochemistry 10-50 μg/mL, Immunohistochemistry-Paraffin 10-50 μg/mL | |
| Q8BWC0 | |
| TPCN2 | |
| A synthetic peptide from amino acid region 700-752 of human Two pore calcium channel protein 2 conjugated to blue carrier protein was used as the antigen. | |
| 0.5 mg | |
| Primary | |
| Human. Other species not yet tested. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction