missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Two pore calcium channel protein 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
176.00€ - 521.00€
Specifications
| Antigen | Two pore calcium channel protein 2 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216293
|
Novus Biologicals
NBP1-30670-0.1MG |
0.1 mg |
176.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18010836
|
Novus Biologicals
NBP1-30670 |
0.5 mg |
521.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Two pore calcium channel protein 2 Polyclonal specifically detects Two pore calcium channel protein 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Two pore calcium channel protein 2 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8BWC0 | |
| 219931 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| Sterilized water with No Preservative | |
| FLJ41094, SHEP10, TPC2two pore calcium channel protein 2, two pore segment channel 2, two-pore calcium channel protein 2, Voltage-dependent calcium channel protein TPC2 | |
| TPCN2 | |
| Primary | |
| Human. Other species not yet tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title