missing translation for 'onlineSavingsMsg'
Learn More

UBE2Q2 Antibody - BSA Free, R&D Systems™

Product Code. 18620512 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620512 0.02 mL 0.02mL
18614062 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18620512 Supplier Novus Biologicals Supplier No. NBP2943500.02ml
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

UBE2Q2 Polyclonal specifically detects UBE2Q2 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2Q2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias DKFZp762C143, EC 6.3.2.19, LOC92912, Ubiquitin carrier protein Q2, ubiquitin-conjugating enzyme E2 Q2, ubiquitin-conjugating enzyme E2Q (putative) 2, ubiquitin-conjugating enzyme E2Q family member 2, Ubiquitin-protein ligase Q2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human UBE2Q2 (NP_775740.1). MSVSGLKAELKFLASIFDKNHERFRIVSWKLDELHCQFLVPQQGSPHSLPPPLTLHCNITESYPSSSPIWFVDSEDPNLTSVLERLEDTKNNNLLRQQLKWLICELCSLYNLPKHLDVEMLDQPLPTGQNGTTEEVTSEEEEEEEEMAED
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 92912
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.