Learn More
Abeba™ UNI6 31752 Shoes
Reflective material provides high visibility. Inner lining is moisture-absorbing, antimicrobial, breathable and hypo-allergenic.
110.16€
Specifications
Certifications/Compliance | EN ISO 20345: S2 |
---|---|
Color | Black, Brown |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Thermoplastic Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15649107
|
Abeba™
31752/35 |
35 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15659107
|
Abeba™
31752/36 |
36 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15669107
|
Abeba™
31752/37 |
37 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15679107
|
Abeba™
31752/38 |
38 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15689107
|
Abeba™
31752/39 |
39 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15699107
|
Abeba™
31752/40 |
40 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15609117
|
Abeba™
31752/41 |
41 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15619117
|
Abeba™
31752/42 |
42 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15629117
|
Abeba™
31752/43 |
43 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15639117
|
Abeba™
31752/44 |
44 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15649117
|
Abeba™
31752/45 |
45 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15659117
|
Abeba™
31752/46 |
46 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15669117
|
Abeba™
31752/47 |
47 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15679117
|
Abeba™
31752/48 |
48 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Safety standard EN ISO 20345: S2
- Color: Black/Brown
- Smooth leather
- Sanitized treated: odor and bacteria inhibiting
- Reflective material for increased visibility
- Interchangeable insole: cushioning, weight distribution, moisture dissipating
- Forefoot is supported and promotes the natural rolling movement of the foot
- Damping PU midsole with joint stabilizer
- Dimensionally stable shaft edge and rear cap
- TPU outsole slip-resistant to SRC
- Moisture grooves run through the sole
- Heel loop for easy entry
- Toe cap made of steel
- ESD capable according to EN 61340
Specifications
EN ISO 20345: S2 | |
Leather | |
Thermoplastic Polyurethane | |
Steel |
Black, Brown | |
Slip-resistant | |
31752 |