missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (RM0143-7J34) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22519
This item is not returnable.
View return policy
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for Western Blot.
Specifications
| uPAR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| P35456 | |
| PLAUR | |
| Mouse recombinant protein of uPAR | |
| 0.1 mg | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0143-7J34 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 5329 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction