missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (RM0143-7J34) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
195.00€ - 507.00€
Specifications
| Antigen | uPAR |
|---|---|
| Clone | RM0143-7J34 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18298064
|
Novus Biologicals
NBP1-22519-0.025MG |
0.025 mg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18167359
|
Novus Biologicals
NBP1-22519 |
0.1 mg | |||||||
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for Western Blot.Specifications
| uPAR | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| PLAUR | |
| IgG2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RM0143-7J34 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P35456 | |
| 5329 | |
| Mouse recombinant protein of uPAR | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title