missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (RM0143-7J34) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22519-0.025MG
This item is not returnable.
View return policy
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for Western Blot.
Specifications
| uPAR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| P35456 | |
| PLAUR | |
| Mouse recombinant protein of uPAR | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0143-7J34 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 5329 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction