missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP17L2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17137478 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137478 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17137478 Supplier Invitrogen™ Supplier No. PA5116241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP17L2 Polyclonal Antibody for Western Blot

DUB3 is a member of the ubiquitin processing protease (UBP) subfamily of deubiquitinating enzymes. See USP1 (MIM 603478) for background information.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L2
Applications Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene USP17L2
Gene Accession No. Q6R6M4
Gene Alias deubiquitinating enzyme 17-like protein 2; deubiquitinating enzyme 3; deubiquitinating protein 3; DUB3; DUB-3; Ubiquitin carboxyl-terminal hydrolase 17; ubiquitin carboxyl-terminal hydrolase 17-like protein 2; ubiquitin specific peptidase 17-like 2; ubiquitin specific peptidase 17-like family member 2; ubiquitin thioesterase 17-like protein 2; ubiquitin thiolesterase 17-like protein 2; ubiquitin-specific-processing protease 17-like protein 2; USP17; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L2; USP17M
Gene Symbols USP17L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 476-503 amino acids from the C-terminal region of human USP17L2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.