missing translation for 'onlineSavingsMsg'
Learn More

Vadimezan, TRC

Product Code. 30455057
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455057 10 mg 10mg
30450487 100 mg -
30455248 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455057 Supplier Toronto Research Chemicals Supplier No. TRCV08495010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117570-53-3
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.29
Synonym 2-(5,6-Dimethyl-9-oxo-9H-xanthen-4-yl)acetic Acid, 5,6-Dimethyl-9-oxo-9H-xanthen-4-ylacetic Acid, 5,6-Dimethylxanthenone-4-acetic Acid, AS 1404, ASA 404, DMXAA, NSC 640488, 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid
IUPAC Name 2-(5,6-dimethyl-9-oxoxanthen-4-yl)acetic acid
SMILES Cc1ccc2C(=O)c3cccc(CC(=O)O)c3Oc2c1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H14O4/c1-9-6-7-13-15(20)12-5-3-4-11(8-14(18)19)17(12)21-16(13)10(9)2/h3-7H,8H2,1-2H3,(H,18,19)
Formula Weight 282.0892
Chemical Name or Material Vadimezan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.