missing translation for 'onlineSavingsMsg'
Learn More

Valproic Acid-d15 Beta-D-Glucuronide, TRC

Product Code. 30457075
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457075 1 mg 1mg
30454279 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457075 Supplier Toronto Research Chemicals Supplier No. TRCV0947631MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valproic Acid-d15 beta-D-Glucuronide
Molecular Formula C14H9D15O8
Quantity 1 mg
Synonym 1-(2-Propylpentanoate-d15) β-D-Glucopyranuronic Acid, 1-O-(Valproyl-d15)-β-D-glucopyranuronic Acid, Dipropylacetate-d15 Glucuronide, VPA-G-d15, Valproate-d15 Glucuronide, Valproic Acid-d15 Glucuronide
InChI Formula InChI=1S/C14H24O8/c1-3-5-7(6-4-2)13(20)22-14-10(17)8(15)9(16)11(21-14)12(18)19/h7-11,14-17H,3-6H2,1-2H3,(H,18,19)/t8-,9-,10+,11-,14-/m0/s1/i1D3,2D3,3D2,4D2,5D2,6D2,7D
SMILES O[C@H]([C@@H](C(O)=O)O[C@@H](OC(C([2H])(C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)[C@@H]1O)[C@@H]1O
Molecular Weight (g/mol) 335.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.