missing translation for 'onlineSavingsMsg'
Learn More

VEGFR1/Flt-1 Antibody (SY09-09), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-11864-100ul

 View more versions of this product

Product Code. 18628727

  • ONLINE EXCLUSIVE PRICE
    549.00€
    518.70€ / 100µL
    Save 30.30€
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VEGFR1/Flt-1
Monoclonal
Unconjugated
1X TBS (pH7.4), 1% BSA, 40% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
SY09-09
Western Blot 1:1000 - 1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry 1:50-1:200
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Recombinant protein (Uniprot#: P17948)
100 μg
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
2321
Store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.