missing translation for 'onlineSavingsMsg'
Learn More

VEGFR1/Flt-1 Antibody (SY09-09), Novus Biologicals™

Rabbit Monoclonal Antibody

253.05€ - 518.70€

Specifications

Antigen VEGFR1/Flt-1
Clone SY09-09
Dilution Western Blot 1:1000 - 1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry 1:50-1:200
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18628727
View Documents
Novus Biologicals
NBP3-11864-100ul
100 μg
ONLINE EXCLUSIVE PRICE
549.00€
518.70€ / 100µL
Save 30.30€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18602168
View Documents
Novus Biologicals
NBP3-11864-25ul
25 μg
ONLINE EXCLUSIVE PRICE
268.00€
253.05€ / 25µL
Save 14.95€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications

Specifications

VEGFR1/Flt-1
Western Blot 1:1000 - 1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Recombinant protein (Uniprot#: P17948)
Protein A purified
SY09-09
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
1X TBS (pH7.4), 1% BSA, 40% Glycerol
2321
Primary
Store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.