missing translation for 'onlineSavingsMsg'
Learn More

Veratridine, TRC

Product Code. 30451393
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451393 1 mg 1mg
30457251 10 mg 10mg
30452893 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451393 Supplier Toronto Research Chemicals Supplier No. TRCV1275681MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-62-5
Molecular Formula C36 H51 N O11
Molecular Weight (g/mol) 673.79
Synonym Cevane-3,4,12,14,16,17,20-heptol, 4,9-epoxy-, 3-(3,4-dimethoxybenzoate), (3β,4α,16β)-, Veratridine (6CI,7CI,8CI), 3-Veratroylveracevine, NSC 7524, Veracevine, veratrate
SMILES COc1ccc(cc1OC)C(=O)O[C@H]2CC[C@@]3(C)[C@@H]4CC[C@H]5[C@]6(O)C[C@H](O)[C@@]7(O)[C@@H](CN8C[C@@H](C)CC[C@H]8[C@@]7(C)O)[C@]6(O)C[C@]35O[C@]24O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C36H51NO11/c1-19-6-11-26-31(3,40)35(43)25(17-37(26)16-19)33(42)18-34-24(32(33,41)15-27(35)38)10-9-23-30(34,2)13-12-28(36(23,44)48-34)47-29(39)20-7-8-21(45-4)22(14-20)46-5/h7-8,14,19,23-28,38,40-44H,6,9-13,15-18H2,1-5H3/t19-,23-,24-,25-,26-,27-,28-,30-,31+,32+,33+,34+,35-,36-/m0/s1
Formula Weight 673.3462
Chemical Name or Material Veratridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.