Learn More
GALR3 Polyclonal Antibody, Invitrogen™
Product Code. 11506212 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11506212 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11506212 Supplier Invitrogen Supplier No. PA120549
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Rabbit Polyclonal Antibody
PA1-20549 detects Galanin Receptor GalR3 in human samples.
The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.
TRUSTED_SUSTAINABILITY
Specifications
Antigen GALR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GALR3
Gene Accession No. O60755, O88626
Gene Alias Gal receptor-3; Gal3 receptor; GAL3-R; Galanin r3 receptor; galanin receptor 3; galanin receptor type 3; galanin receptor, family member 3; Galnr3; Galr3; Gal-r3; GALR-3; OTTHUMP00000028820
Gene Symbols GALR3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal domain of human Galanin Receptor GalR3.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29235, 8484
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG