Learn More
Invitrogen™ STK35 Polyclonal Antibody
GREENER_CHOICE
Product Code. 17198363
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17198363 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17198363 Supplier Invitrogen™ Supplier No. PA5110441
Rabbit Polyclonal Antibody
Immunogen sequence: RPEGGGGSAR PRYSLLAEIG RGSYGVVYEA VAGRSGARVA VKKIRCDAPE NVELALAEFW ALTSLKRRHQ NVVQFEECVL QRNGLAQRMS HGNKSSQLYL RLVETSLKGE RILGYAEEPC YLWFVMEFCE GGDLNQYVLS RRPDPATNKS FMLQLTSAIA FLHKNHIVHR DLKPDNILIT ERSGTPILKV A.
The protein encoded by this gene is a kinase that is predominantly found in the nucleus. However, it can interact with PDLIM1/CLP-36 in the cytoplasm and localize to actin stress fibers. The encoded protein may be a regulator of actin stress fibers in nonmuscle cells.
TRUSTED_SUSTAINABILITY
Specifications
Antigen STK35
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Stk35
Gene Accession No. Q80ZW0
Gene Alias 1700054C12Rik; bA550O8.2; bA550O8.2 CLIK1; CLIK1; CLIK-1; CLP-36 interacting kinase; CLP-36-interacting kinase 1; OTTHUMP00000030006; PDIK1; PDLIM1-interacting kinase 1; serine threonine kinase 35 long form; serine/threonine kinase 35; serine/threonine-protein kinase 35; Serine/threonine-protein kinase 35 L1; STK35; Stk35l1
Gene Symbols Stk35
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 190-380 of human STK35 (NP_543026.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67333
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG