Description
TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
Specifications
| Antigen | TAFA3/FAM19A3 |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), CyTOF |
| Classification | Monoclonal |
| Clone | 460904 |
| Conjugate | DyLight 594 |
| Dilution | Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8 |
| Show More |