Description
DOCK11 Polyclonal antibody specifically detects DOCK11 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
Specifications
| Antigen | DOCK11 |
| Applications | Immunoprecipitation, Western Blot (Negative) |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | ACG, Activated Cdc42-associated guanine nucleotide exchange factor, Activated Cdc42-associated guanine nucleotide exchange factor, ACG, bB128O4.1, bB128O4.1.1 (novel protein), dedicator of cytokinesis 11, FLJ32122, FLJ43653, ZIZ2dedicator of cytokinesis protein 11, zizimin2, zizimin-2 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human dedicator of cytokinesis 11 using the numbering given in entry NP_653259.3 (GeneID 139818). |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |