missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VKORC1 Polyclonal antibody specifically detects VKORC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | VKORC1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.1.4.1, EDTP308, FLJ00289, IMAGE3455200, MST576, phylloquinone epoxide reductase, vitamin K dependent clMST134, vitamin K epoxide reductase complex subunit 1, vitamin K epoxide reductase complex, subunit 1, Vitamin K1 2,3-epoxide reductase subunit 1, vitamin K1 epoxide reductase (warfarin-sensitive), VKCFD2, VKORMGC2694 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human VKORC1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?