missing translation for 'onlineSavingsMsg'
Learn More

Voclosporin-d4, TRC

Product Code. 30457171
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457171 5 mg 5mg
30450597 1 mg 1mg
30456082 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457171 Supplier Toronto Research Chemicals Supplier No. TRCV6785025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Voclosporin-d4
Molecular Formula C63H107D4N11O12
Quantity 5 mg
Synonym 6-[(2S,3R,4R,6E)-3-Hydroxy-4-methyl-2-(methylamino)-6,8-nonadienoic acid]cyclosporin A-d4, ISATX 247-d4, R 1524-d4, trans-ISA 247-d4
InChI Formula InChI=1S/C63H111N11O12/c1-25-27-28-29-41(15)53(76)52-57(80)66-44(26-2)59(82)68(18)34-49(75)69(19)45(30-35(3)4)56(79)67-50(39(11)12)62(85)70(20)46(31-36(5)6)55(78)64-42(16)54(77)65-43(17)58(81)71(21)47(32-37(7)8)60(83)72(22)48(33-38(9)10)61(84)73(23)51(40(13)14)63(86)74(52)24/h25,27-28,35-48,50-53,76H,1,26,29-34H2,2-24H3,(H,64,78)(H,65,77)(H,66,80)(H,67,79)/b28-27+/t41-,42+,43-,44+,45+,46+,47+,48+,50+,51+,52+,53-/m1/s1/i1D2,25D,27D
SMILES CC[C@H]1C(N(C)CC(N(C)[C@@H](CC(C)C)C(N[C@@H](C(C)C)C(N(C)[C@@H](CC(C)C)C(N[C@@H](C)C(N[C@H](C)C(N(C)[C@@H](CC(C)C)C(N(C)[C@@H](CC(C)C)C(N(C)[C@@H](C(C)C)C(N(C)[C@@H]([C@H](O)[C@@H](C/C=C([2H])/C([2H])=C([2H])/[2H])C)C(N1)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-ethyl-33-((1R,2R,E)-1-hydroxy-2-methylhepta-4,6-dien-1-yl-5,6,7,7-d4)-6,9,18,24-tetraisobutyl-3,21-diisopropyl-1,4,7,10,12,15,19,25,28-nonamethyl-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecaone
Molecular Weight (g/mol) 1218.65
Formula Weight 1217.87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.