missing translation for 'onlineSavingsMsg'
Learn More

von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Product Code. 12632037 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
Unit Size:
100µL
1mL
200µL
500µL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Concentration unitSize
12632037 100 μL Biotin 200μg/mL 100µL
12642037 500 μL Biotin 200μg/mL 500µL
12662037 100 μL Unconjugated 200μg/mL 100µL
12672037 500 μL Unconjugated 200μg/mL 500µL
12652037 1 mL Unconjugated 200μg/mL 1mL
12682037 100 μL Unconjugated 1mg/mL 100µL
12692037 200 μL Unconjugated 1mg/mL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12632037 Supplier Epredia Supplier No. MS690B0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide accurate, reproducible results with the Epredia™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody.

Molecular genetic studies suggest that VHL is a tumor-suppressor gene. Alterations in the von Hippel-Lindau (VHL) gene are correlated with a diverse group of neoplasms including hemangioblastoma, clear cell renal carcinoma (RCC), and pheochromocytoma. Two transcriptional elongation factors, Elongin B and C, are shown to bind in vitro and in vivo to a short, colinear region of the VHL protein (pVHL) that is frequently mutated in human tumors. It is proposed that pVHL functions in vivo as a negative regulator of transcription elongation.

Host Species: Mouse

Clone: Ig33

Isotype: IgG1/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: aa 1-54

Immunogen: A recombinant protein containing aa 1-213 of human vHL

Molecular Weight: 30kDa

Positive Control: Kidney

Cellular Localization: Cytoplasmic Ab-1 does not recognize the truncated form of VHL protein.

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen von Hippel-Lindau (vHL) Protein Ab-1
Classification Monoclonal
Clone Ig33
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A recombinant protein containing aa 1-213 of human VHL
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.