missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VSTM1 Monoclonal Antibody (1A5)
GREENER_CHOICE

Product Code. 17195263
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17195263 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17195263 Supplier Invitrogen™ Supplier No. MA533372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Application Notes: Western blot - non-reduced sample treatment and SDS-Page was used. The band sizes are 45 kDa and fainter band of 37 kDa in transfected cells and 37 kDa in granulocytes. Functional Assay - Antibody 1A5 showed inhibition of Fc eta RI-mediated degranulation by RBL-2H3 cells transfected with SIRL-1. The percentage of inhibition of degranulation by SIRL-1 was calculated by (OD405 TNP stimulation - OD405 TNP with SIRL-1 crosslinking)/(OD405 TNP stimulation - OD405 spontaneous release) x 100.

VSTM1 (V-Set And Transmembrane Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is LILRB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSTM1
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A5
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene VSTM1
Gene Accession No. Q6UX27
Gene Alias LAIR homolog; OSCAR-like transcript-1; signal inhibitory receptor on leukocytes-1; SIRL1; SIRL-1; UNQ3033; UNQ3033/PRO9835; V-set and transmembrane domain containing 1; V-set and transmembrane domain-containing protein 1; VSTM1
Gene Symbols VSTM1
Host Species Mouse
Immunogen Purified SIRL-1-hIg fusion protein in PBS.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284415
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.